missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin A/Lysosomal Carboxypeptidase A Antibody, R&D Systems™

Description
Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
| Antigen | Cathepsin A/Lysosomal Carboxypeptidase A |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 179803 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P10619 |
| Gene Alias | beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, CTSA, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, Lysosomal Carboxypeptidase A, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?