missing translation for 'onlineSavingsMsg'
Learn More

Human Cathepsin D Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

214.00 € - 403.00 €

Specifications

Antigen Cathepsin D
Clone 984824
Dilution Immunocytochemistry 8-25 ug/mL
Applications Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18738189
GREENER_CHOICE
View Documents
R&D Systems
MAB10141-SP
25 μg
214.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18700979
GREENER_CHOICE
View Documents
R&D Systems
MAB10141-100
100 μg
403.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cathepsin D Monoclonal specifically detects Cathepsin D in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

Cathepsin D
Immunocytochemistry 8-25 ug/mL
Monoclonal
Supernatant
Human
P07339
1509
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
984824
Immunocytochemistry
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Mouse myeloma cell line NS0-derived recombinant human Cathepsin D Leu21-Leu412 Accession # P07339
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Cathepsin D in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.