missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin L Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB952-SP
This item is not returnable.
View return policy
Description
Cathepsin L Monoclonal specifically detects Cathepsin L in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cathepsin L | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cathepsin L, cathepsin L1, CATL, CTSL, CTSL1, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1514 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 204106 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P07711 | |
| CTSL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin L Glu113-Val333 Accession # P07711 | |
| 25 μg | |
| Primary | |
| Detects human Cathepsin L in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Cathepsin B, C, L2, O, S, X/Z/P, or recombinant mouse Cathepsin H and L is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction