missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL11/Eotaxin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3201
This item is not returnable.
View return policy
Description
CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| CCL11/Eotaxin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL11 | |
| E. coli-derived recombinant human CCL11/Eotaxin | |
| 500 μg | |
| Primary | |
| Detects human CCL11/Eotaxin in Western blots. Does not cross-react with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 3, 4, 5, 8, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, or recombinant rat CCL20. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 43915 | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6356 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction