missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL11/Eotaxin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
213.00 € - 2050.00 €
Specifications
| Antigen | CCL11/Eotaxin |
|---|---|
| Clone | 43911 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 1-5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702949
![]() |
R&D Systems
MAB320-SP |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18724378
![]() |
R&D Systems
MAB320-100 |
100 μg |
519.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711387
![]() |
R&D Systems
MAB320-500 |
500 μg |
2050.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CCL11/Eotaxin | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 1-5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| CCL11 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 43911 | |
| Western Blot, Immunohistochemistry, Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P51671 | |
| 6356 | |
| E. coli-derived recombinant human CCL11/Eotaxin Gly24-Pro97 Accession # P51671 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CCL11/Eotaxin in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 9/10/MIP-1 gamma , 14, 17, 19, 20, 21, 25, recombinant mouse CCL2, 3 4 56 7, 9/10/MIP-1 gamma , 11, 21, or 25. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title