missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 369.00 €
Specifications
| Antigen | CCR1 |
|---|---|
| Clone | 984303 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746328
![]() |
R&D Systems
MAB1451-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752059
![]() |
R&D Systems
MAB1451-100 |
100 μg |
369.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR1 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P32246 | |
| 1230 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 984303 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| Mouse myeloma cell line NS0-derived recombinant human CCR1 Met1-Phe355 Accession # P32246 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CCR1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title