missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR6 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems FAB195C-025
This item is not returnable.
View return policy
Description
CCR6 Monoclonal specifically detects CCR6 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR6 | |
| Monoclonal | |
| PerCP | |
| P51684 | |
| CCR6 | |
| NS0 mouse myeloma cell line transfected with human CCR6 Met1-Met374 Accession # P51684 | |
| 25 Tests | |
| 1235 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 53103 | |
| Flow Cytometry 10 uL/10^6 cells | |
| BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human CCR6. Stains human CCR6 transfectants but not the parental cell line in flow cytometry. Does not cross-react with transfectants expressing human CCR1, CCR2, CCR3, CCR4, CCR5, CCR7, CXCR1, CXCR2, CXCR3, CXCR4, or STRL33. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction