missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR9 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
201.00 € - 396.00 €
Specifications
| Antigen | CCR9 |
|---|---|
| Clone | 112509 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794707
![]() |
R&D Systems
FAB179F-025 |
25 Tests |
201.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766286
![]() |
R&D Systems
FAB179F-100 |
100 Tests |
396.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR9 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| CCR9 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Specifically stains human CCR9 transfectants and MOLT-4 cells. It does not react with the parental BaF3 cells or human CCR7-transfected BaF3 cells. |
| 112509 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| P51686 | |
| 10803 | |
| BaF3 mouse pro-B cell line transfected with human CCR9 Met1-Leu369 Accession # P51686 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title