missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD11c PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
252.00 € - 458.00 €
Specifications
| Antigen | CD11c |
|---|---|
| Clone | ICRF 3.9 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781178
![]() |
R&D Systems
FAB1777P-025 |
25 Tests |
252.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18736476
![]() |
R&D Systems
FAB1777P-100 |
100 Tests |
458.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for Flow Cytometry.Specifications
| CD11c | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 3687 | |
| Rheumatoid synovial fluid cells and human monocyte-derived fibronectin | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| ICRF 3.9 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, Integrin alpha X, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), ITGAX, Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, p150,95 alpha, SLEB6 | |
| ITGAX | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD11c in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title