missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD14 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
209.00 € - 1735.00 €
Specifications
| Antigen | CD14 |
|---|---|
| Clone | 50040 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772549
![]() |
R&D Systems
MAB3833-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18750078
![]() |
R&D Systems
MAB3833-100 |
100 μg |
428.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18794269
![]() |
R&D Systems
MAB3833-500 |
500 μg |
1735.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| CD14 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P08571 | |
| 929 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD14 in sandwich ELISAs. |
| 50040 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| CD14 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title