missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human CD155/PVR Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
228.00 € - 537.00 €
Specifications
| Antigen | CD155/PVR |
|---|---|
| Clone | 1072806 |
| Dilution | Flow Cytometry 2.5 μg/106 cells, Immunocytochemistry 3-25 μg/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018978
![]() |
Novus Biologicals™
MAB11503-100 |
100 μg |
537.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019022
![]() |
Novus Biologicals™
MAB11503-SP |
25 μg |
228.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Flow Cytometry,ImmunocytochemistrySpecifications
| CD155/PVR | |
| Flow Cytometry 2.5 μg/106 cells, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4 | |
| Mouse myeloma cell line NS0-derived recombinant human CD155/PVR, Gly27-Asn343, Accession # P15151 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1072806 | |
| Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P15151 | |
| 5817 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title