missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD25/IL-2R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB223-500
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry.
Specifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3559 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry | |
| 22722 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.5-1.0 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P01589 | |
| IL2RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD25/IL-2 R alpha Glu22-Cys213 Accession # P01589 | |
| 500 μg | |
| Primary | |
| Detects human CD25/IL-2 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 4% cross-reactivity with recombinant human (rh) IL-2 R beta is observed and no cross-reactivity with rhIL-2 R gamma , rhIL-1 RI, rhIL-1 RII, rhIL-4 R, rhIL-5 R alpha , rhIL-6 R, rhIL-7 R, rhIL-9 R, and rhIL-10 R is observed. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction