missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD27/TNFRSF7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 2040.00 €
Specifications
| Antigen | CD27/TNFRSF7 |
|---|---|
| Clone | 57703 |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.2-0.4 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711458
![]() |
R&D Systems
MAB382-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718528
![]() |
R&D Systems
MAB382-100 |
100 μg |
517.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18717679
![]() |
R&D Systems
MAB382-500 |
500 μg |
2040.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.Specifications
| CD27/TNFRSF7 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.2-0.4 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| P26842 | |
| 939 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD27 Thr21-Ile192 Accession # P26842 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD27 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, recombinant mouse (rm) CD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhLTR beta , rhNGF R, rhOPG, rmOX40, rhRANK, rhTAJ, or rhTNF RI is observed. |
| 57703 | |
| Western Blot, Flow Cytometry, Neutralization, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD27, CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
| CD27 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title