missing translation for 'onlineSavingsMsg'
Learn More

Human CD3 delta/CD3d Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

214.00 € - 448.00 €

Specifications

Antigen CD3 delta
Clone 1033614
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18762797
GREENER_CHOICE
View Documents
R&D Systems
MAB10672-SP
25 μg
214.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18703167
View Documents
R&D Systems
MAB10672-100
100 μg
448.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD3 delta Monoclonal antibody specifically detects CD3 delta in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

CD3 delta
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
CD3 antigen, delta subunit, CD3D, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
Human embryonic kidney cell HEK293-derived human CD3 delta/CD3d, Phe22-Ala105, Accession # P04234
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1033614
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Human
P04234
915
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.