missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD3 epsilon Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB100V-100
This item is not returnable.
View return policy
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry.
Specifications
| CD3 epsilon | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD3E | |
| Human thymocytes (1) | |
| 100 Tests | |
| Primary | |
| Recognizes the epsilon-chain of the CD3/T cell antigen receptor complex (McMichael, A.J. et al. (1987) Leucocyte Typing III: White Cell Differentiation Antigens, Oxford University Press, New York; Knapp, W. et al. (1989) Leucocyte Typing IV: White Cell Differentiation Antigens, Oxford University Press, New York; Schlossman, S. et al. (1995) Leucocyte Typing V: White Cell Differentiation Antigens, Oxford University Press, New York). | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| UCHT1 | |
| Flow Cytometry 5 uL/10^6 cells | |
| CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 916 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction