missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD30 Ligand/TNFSF8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1028-500
This item is not returnable.
View return policy
Description
CD30 Ligand/TNFSF8 Monoclonal specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.
Specifications
| CD30 Ligand/TNFSF8 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-4 ug/mL, CyTOF-ready | |
| P32971 | |
| TNFSF8 | |
| Mouse myeloma cell line NS0-derived recombinant human CD30 Ligand/TNFSF8 Gln63-Asp234 Accession # P32971 | |
| 500 μg | |
| Primary | |
| Detects human CD30 Ligand/TNFSF8 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CD27 Ligand, recombinant mouse CD30 Ligand, or rhCD40 Ligand is observed. | |
| Human | |
| Supernatant |
| Flow Cytometry, Neutralization, CyTOF | |
| 116614 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD153, CD153 antigen, CD30 antigen ligand, CD30L, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, TNFSF8, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 944 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction