missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD30/TNFRSF8 F(ab')2 (Research Grade Brentuximab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
218.00 € - 601.00 €
Specifications
| Antigen | CD30/TNFRSF8 |
|---|---|
| Clone | Hu8F2 |
| Dilution | Flow Cytometry 0.25 ug/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755755
![]() |
R&D Systems
MAB11129-FAB2-100 |
100 μg |
601.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18765815
![]() |
R&D Systems
MAB11129-FAB2-SP |
25 μg |
218.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow CytometrySpecifications
| CD30/TNFRSF8 | |
| Flow Cytometry 0.25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 943 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Hu8F2 | |
| Flow Cytometry | |
| Unconjugated | |
| Human | |
| Human | |
| CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 | |
| Human CD30/TNFRSF8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title