missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD300a/LMIR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
207.00 € - 548.00 €
Specifications
| Antigen | CD300a/LMIR1 |
|---|---|
| Clone | 232618 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755905
![]() |
R&D Systems
MAB26401-100 |
100 μg |
548.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766095
![]() |
R&D Systems
MAB26401-SP |
25 μg |
207.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Human samples. It is validated for ELISASpecifications
| CD300a/LMIR1 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 11314 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 232618 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Human | |
| CD300a, CD300a antigenCMRF35H9, CD300a molecule, CLM8, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, IRp60, leukocyte membrane antigen, LMIR1, MAIR-I, Mcipr1, NKRL, Pigr4 | |
| Y3 rat myeloid cell line transfected with human CD300a/LMIR1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title