missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD300f/LMIR3 Antibody, R&D Systems™
Description
CD300f/LMIR3/CD300LF Polyclonal specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | CD300f/LMIR3/CD300LF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL |
| Gene Accession No. | Q8TDQ1 |
| Gene Alias | CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CD300LF, CLIM1, CLM1, CLM-1, DIgR2, IgSF13, IREM1, IREM-1, LMIR3, NKIR, Pigr3 |
| Gene Symbols | CD300LF |
| Host Species | Goat |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?