Learn More
Human CD31/PECAM-1 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11666-SP
 View more versions of this product
Product Code. 30306746
  • 247.00 € / 25µg
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD31/PECAM-1
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Chinese Hamster Ovary cell line, CHO-derived recombinant human CD31/PECAM01, Gln28-Lys601, Accession # P16284
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Purified
Western Blot, Simple Western, Immunohistochemistry, Multiplex Immunofluorescence
1094431
Western Blot 2 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL
P16284
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5175
Human
Antibody
IgG2b