Learn More
Human CD31/PECAM-1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems FAB3567A-100
 View more versions of this product
Product Code. 18758476
  • 462.00 € / 100 tests
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Flow Cytometry, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD31/PECAM-1
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
PECAM1
Activated HUVEC human umbilical vein endothelial cells
100 Tests
Primary
Detects human CD31/PECAM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) E-Selectin, rhICAM-1, -2, -3, rhVCAM-1, or recombinant mouse VCAM-1 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry, Immunoassay
9G11
Flow Cytometry 10 μL/10^6 cells, Knockout Validated
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5175
Human
Purified