Learn More
Human CD34 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB72274-SP
 View more versions of this product
Product Code. 18132521
  • 221.00 € / 25µg
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD34
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1
Chinese Hamster Ovary cell line, CHO-derived human CD34 protein, Ser32-Thr290, Accession # P28906-1
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG2b
Flow Cytometry, Immunocytochemistry
1054015
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
P28906-1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
947
Human
Purified