missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD45RO Antibody, R&D Systems™

Mouse Monoclonal Antibody
204.00 € - 429.00 €
Specifications
| Antigen | CD45RO |
|---|---|
| Clone | UCHL1 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794317
![]() |
R&D Systems
MAB10642-SP |
25 μg |
204.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783697
![]() |
R&D Systems
MAB10642-100 |
100 μg |
429.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow CytometrySpecifications
| CD45RO | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 5788 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| UCHL1 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Cultured T cells from an IL-2-dependent T-cell line (CA1) prepared from human peripheral blood activated with influenza virus. | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title