missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD8 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3801-SP
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Immunohistochemistry.
Specifications
| CD8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD8, CD8A, CD8a molecule, LEU2, p32 | |
| Mouse myeloma cell line NS0-derived recombinant human CD8 alpha , Ser22-Val198, Accession No. NP_741696 | |
| 25 μg | |
| 925.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 37020 | |
| Immunohistochemistry 5-25 μg/mL | |
| NP_741696 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CD8 alpha in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction