Learn More
Human CEACAM-16 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB9885-100
 View more versions of this product
Product Code. 18752447
  • 480.00 € / 100µg
Description
CEACAM16 Monoclonal antibody specifically detects CEACAM16 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEACAM16
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
carcinoembryonic antigen-related cell adhesion molecule 16, CEAL2, DFNA4B
Chinese Hamster Ovary cell line CHO-derived human CEACAM-16, Met1-Gly425, Accession # Q2WEN9
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Flow Cytometry, Immunocytochemistry
2747B
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL
Q2WEN9
Rabbit
Protein A or G purified from cell culture supernatant
RUO
388551