missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-18 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9869R-100UG
This item is not returnable.
View return policy
Description
CEACAM18 Monoclonal antibody specifically detects CEACAM18 in Human samples. It is validated for Flow Cytometry
Specifications
| CEACAM18 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Carcinoembryonic Antigen-Related Cell Adhesion Molecule 18 | |
| Human embryonic kidney cell, HEK293-derived human CEACAM-18, Gln31-His317, Accession # A8MTB9 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1057633 | |
| Flow Cytometry | |
| A8MTB9 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 729767 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction