Learn More
Human CEACAM-19 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB93971N-100UG
Product Code. 18700117
  • 449.00 € / 100µg
Description
CEACAM19 Monoclonal antibody specifically detects CEACAM19 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEACAM19
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
carcinoembryonic antigen-related cell adhesion molecule 19, CEACAM19, CEAL1
Chinese hamster ovary cell line CHO-derived recombinant human CEACAM-19, Met1-Gly157, Accession # Q7Z692
100 μg
Primary
Human
Purified
Flow Cytometry
1042108
Flow Cytometry
Q7Z692
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
56971
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
IgG1