missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-3/CD66d Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems BAF4166
This item is not returnable.
View return policy
Description
CEACAM3/CD66d Polyclonal specifically detects CEACAM3/CD66d in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CEACAM3/CD66d | |
| Polyclonal | |
| Biotin | |
| P40198 | |
| CEACAM3 | |
| Mouse myeloma cell line NS0-derived recombinant human CEACAM-3/CD66d isoform 1 Lys35-Gly155 Accession # P40198 | |
| 50 μg | |
| Primary | |
| Detects human CEACAM-3/CD66d in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human (rh) CEACAM-1 is observed, 10% cross-reactivity with rhCEACAM-6 is observed, and 5% cross-reactivity with rhCEACAM-4 and rhCEACAM-5 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| CD66D, CEA, CEACAM3 carcinoembryonic antigen-related cell adhesion molecule 3, CGM1, W264, W282 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1084 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction