missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4354
This item is not returnable.
View return policy
Description
CEACAM4 Monoclonal specifically detects CEACAM4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CEACAM4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O75871 | |
| CEACAM4 | |
| E. coli-derived recombinant human CEACAM-4 Phe36-Val140 Accession # O75871 | |
| 100 μg | |
| Primary | |
| Detects human CEACAM-4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CEACAM-1, -3, -5, -6, -7, or -8 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 568410 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| carcinoembryonic antigen CGM7, carcinoembryonic antigen gene family member 7, carcinoembryonic antigen-related cell adhesion molecule 4, CEACAM4, CGM7, CGM7_HUMAN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1089 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction