Learn More
Human CFL1 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11669-100
 View more versions of this product
Product Code. 30484807
  • 572.00 € / 100µg
Description
Cofilin Monoclonal antibody specifically detects Cofilin in Human samples. It is validated for Simple Western,Western Blot,Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cofilin
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
18 kDa phosphoprotein, CFL, cofilin, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, HEL-S-15, p18
Synthetic Peptide, Accession # P23528
100 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Purified
Simple Western, Western Blot, Immunohistochemistry, Immunocytochemistry
1093310
Western Blot 0.5 μg/mL, Simple Western 10-100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
P23528
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
1072
Human
Antibody
IgG2b