missing translation for 'onlineSavingsMsg'
Learn More

Human Chitotriosidase/CHIT1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18708399
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18708399 25 μg 25µg
18152441 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18708399 Supplier R&D Systems Supplier No. MAB35591SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chitotriosidase/CHIT1 Monoclonal specifically detects Chitotriosidase/CHIT1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chitotriosidase/CHIT1
Applications Western Blot
Classification Monoclonal
Clone 397513
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13231
Gene Alias CHI3, CHIT1, CHITCHI3, chitinase 1 (chitotriosidase), Chitinase-1, chitotriosidase-1, EC 3.2.1.14, FLJ00314, MGC125322, plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase
Gene Symbols CHIT1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chitotriosidase/CHIT1 Ala22-Asn466 Accession # Q13231
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1118
Test Specificity Detects human Chitotriosidase/CHIT1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CHI3-L1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG3
Afficher plus Afficher moins

For Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.