missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chk2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
208.00 € - 468.00 €
Specifications
| Antigen | Chk2 |
|---|---|
| Clone | 2521A |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734687
![]() |
R&D Systems
MAB1358-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760307
![]() |
R&D Systems
MAB1358-100 |
100 μg |
468.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chk2 Monoclonal specifically detects Chk2 in Human samples. It is validated for Western Blot.Specifications
| Chk2 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CDS1, CHEK2, CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, HuCds1, LFS2, PP1425, Rad53, S.pombe) homolog | |
| E. coli-derived human Chk2, Asp208-Leu543, Accession # O96017 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2521A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O96017 | |
| 11200 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title