missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CHL-1/L1CAM-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
214.00 € - 443.00 €
Specifications
| Antigen | CHL-1/L1CAM-2 |
|---|---|
| Clone | 316223 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1.7-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18703328
![]() |
R&D Systems
MAB2126-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159780
|
R&D Systems
MAB2126 |
100 μg |
443.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHL-1/L1CAM-2 Monoclonal specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CHL-1/L1CAM-2 | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| NP_006605 | |
| 10752 | |
| Mouse myeloma cell line NS0-derived recombinant human CHL-1/L1CAM-2 Ile25-Gln1096 Accession # NP_006605 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CHL-1 is observed. |
| 316223 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1.7-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
| CHL1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title