missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CHRM5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10323-SP
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Muscarinic Acetylcholine Receptor M5/CHRM5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| cholinergic receptor, muscarinic 5, HM5, M5, M5R, mAChR M5, MGC41838, muscarinic acetylcholine receptor M5 | |
| NS0 mouse myeloma cell line transfected with human CHRM5, Met1-Pro532, Accession # P08912 | |
| 25 μg | |
| Primary | |
| Detects human CHRM5 in diect ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 590123 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P08912 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 1133 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction