missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chromogranin A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB90982-SP
This item is not returnable.
View return policy
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human samples. It is validated for ELISA
Specifications
| Chromogranin A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Betagranin, betagranin (N-terminal fragment of chromogranin A), Catestatin, CGA, CHGA, Chromofungin, chromogranin A (parathyroid secretory protein 1), chromogranin-A, Pancreastatin, Parastatin, parathyroid secretory protein 1, Pituitary Secretory Protein I, SP-I | |
| Human embryonic kidney cell line HEK293-derived human Chromogranin A, Met1-Gly457, Accession # P10645 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 948130 | |
| ELISA | |
| P10645 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1113 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction