missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Claudin-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody
249.00 € - 600.00 €
Specifications
| Antigen | Claudin-11 |
|---|---|
| Clone | 398205R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772547
![]() |
R&D Systems
MAB4280R-SP |
25 μg |
249.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744367
![]() |
R&D Systems
MAB4280R-100 |
100 μg |
600.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin-11 Monoclonal specifically detects Claudin-11 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| Claudin-11 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| claudin 11, claudin-11, Claudin11, CLDN11, Oligodendrocyte-specific protein, OSP, OSPOTMoligodendrocyte transmembrane protein, OTM | |
| NS0 mouse myeloma cell line transfected with human Claudin-11, Met1-Val207, Accession # O75508 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 398205R | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75508 | |
| 5010 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title