missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Collagen XIII alpha 1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6346
This item is not returnable.
View return policy
Description
Collagen XIII alpha 1 Monoclonal specifically detects Collagen XIII alpha 1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Collagen XIII alpha 1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q5TAT6 | |
| COL13A1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Collagen XIII alpha 1 Glu109-Pro668 Accession # Q5TAT6 | |
| 100 μg | |
| Primary | |
| Detects human Collagen XIII alpha 1 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse Collagen XIII alpha 1 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 584718 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COL13A1, collagen alpha-1(XIII) chain, collagen, type XIII, alpha 1, FLJ42485 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1305 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction