missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C1s Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2060
This item is not returnable.
View return policy
Description
Complement Component C1s Polyclonal specifically detects Complement Component C1s in Human samples. It is validated for Western Blot.
Specifications
| Complement Component C1s | |
| Polyclonal | |
| Unconjugated | |
| P09871 | |
| C1S | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Component C1s Glu16-Asp688 Accession # P09871 | |
| 100 μg | |
| Primary | |
| Detects human Complement Component C1s in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human C1r is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| basic proline-rich peptide IB-1, C1 esterase, C1S, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 716 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction