missing translation for 'onlineSavingsMsg'
Learn More

Human Complement Factor H-related 1/CFHR1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18767417
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767417 25 μg 25µg
18058271 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18767417 Supplier R&D Systems Supplier No. AF4247SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Complement Factor H-related 1/CFHR1/CFHL1 Polyclonal specifically detects Complement Factor H-related 1/CFHR1/CFHL1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor H-related 1/CFHR1/CFHL1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q03591
Gene Alias BTA, CFHL1FHR-1, CFHLCFHR1P, CFHR1, complement factor H-related 1, Complement Factor Hrelated 1, complement factor H-related 1 pseudogene, complement factor H-related protein 1, FHR1, FHR1CFHL1P, H factor (complement)-like 1, H factor (complement)-like 2, H factor-like protein 1, H36-1, H36-2, H-factor-like 1, HFL1H36, HFL2, MGC104329
Gene Symbols CFHR1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 1/CFHR1 Glu19-Ala328 Accession # Q03591
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3078
Test Specificity Detects human Complement Factor H-related 1/CFHR1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) FHR2 is observed and approximately 15% cross-reactivity with rhFHR5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.