Learn More
Human CRLR Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB10044-100
 View more versions of this product
Product Code. 18791797
  • 469.00 € / 100µg
Description
CRLR Monoclonal specifically detects CRLR in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CRLR
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
calcitonin gene-related peptide type 1 receptor, calcitonin receptor-like, CALCRL, CGRP type 1 receptor, CGRPR, CGRPRCRLRCalcitonin receptor-like receptor
Synthetic peptide containing human CALCRL, Accession No. Q16602
100 μg
10203.0
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a
Immunohistochemistry, Immunocytochemistry
998820
Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL
Q16602
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human CALCRL in direct ELISAs.
Human
Supernatant