missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTRP3/C1qTNF3/CORS26 Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 468.00 €
Specifications
| Antigen | C1qTNF3/CORS26/CTRP3 |
|---|---|
| Clone | 860101 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793617
![]() |
R&D Systems
MAB79251-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18714007
![]() |
R&D Systems
MAB79251-100 |
100 μg |
468.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C1qTNF3/CORS26/CTRP3 Monoclonal specifically detects C1qTNF3/CORS26/CTRP3 in Human samples. It is validated for ELISA.Specifications
| C1qTNF3/CORS26/CTRP3 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| C1ATNF3, C1q and tumor necrosis factor related protein 3, C1qTNF3, Cartducin, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26 | |
| Mouse myeloma cell line, NS0-derived human CTRP3/C1qTNF3/CORS26, Gln23-Lys246, Accession # Q9BXJ4-1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 860101 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BXJ4-1 | |
| 114899 | |
| IgG3 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title