missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL1/2/3/GRO Pan Specific Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB276-SP
This item is not returnable.
View return policy
Description
CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| CXCL1/2/3/GRO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CINC1, CINC-1, CINC-2, CINC3, CINC-3, Dcip1, FSP, Gm1960, GRO alpha, GRO beta, GRO gamma, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, MGSA-beta, MIP-2, NAP-3, SCYB1 | |
| E. coli-derived recombinant human CXCL2 Ala35-Asn107 Accession # P19875 | |
| 25 μg | |
| Primary | |
| Detects human CXCL1/2/3 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| 31716 | |
| Western Blot 1 ug/mL, Neutralization | |
| P19875 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2919 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto