Learn More
Human CXCL9/MIG Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF392-SP
Product Code. 18769257
  • 209.00 € / 25µg
Description
CXCL9/MIG Polyclonal specifically detects CXCL9/MIG in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL9/MIG
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Goat
Affinity Purified
RUO
4283
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Neutralization, Immunocytochemistry
LYOPH
Western Blot 0.1 ug/mL, Neutralization 3-12 ug/mL, Immunocytochemistry 5-15 ug/mL
Q07325
CXCL9
E. coli-derived recombinant human CXCL9/MIG Thr23-Thr125 Accession # Q07325
25 μg
Primary
Detects human CXCL9/MIG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant mouse CXCL9 (non-reducing conditions) is observed.