missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cyclin D1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB43142-SP
This item is not returnable.
View return policy
Description
Cyclin D1 Monoclonal specifically detects Cyclin D1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Cyclin D1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, CCND1, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), Cyl-1, G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
| E.coli-derived recombinant human Cyclin D1, Met1-Ile295, Accession No. P24385 | |
| 25 μg | |
| 595.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 328311 | |
| Immunocytochemistry 8-25 μg/mL | |
| P24385 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Cyclin D1 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction