missing translation for 'onlineSavingsMsg'
Learn More

Human/Cynomolgus Monkey B7-H6 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18792387
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18792387 25 μg 25µg
18794357 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792387 Supplier R&D Systems Supplier No. MAB10162SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

B7-H6 Monoclonal specifically detects B7-H6 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-H6
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1002910
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived Cynomolgus Monkey B7-H6, Asp25-Ser262
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 374383
Test Specificity Detects human and cynomolgus monkey B7-H6 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Cynomolgus Monkey
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.