missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin C Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
209.00 € - 1820.00 €
Specifications
| Antigen | Cystatin C |
|---|---|
| Clone | 197807 |
| Concentration | LYOPH |
| Dilution | Western Blot 1-2 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18790057
![]() |
R&D Systems
MAB1196-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758879
![]() |
R&D Systems
MAB1196-100 |
100 μg |
452.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741229
![]() |
R&D Systems
MAB1196-500 |
500 μg |
1820.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Cystatin C | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin | |
| CST3 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 197807 | |
| Western Blot 1-2 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01034 | |
| 1471 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin C Ser27-Ala146 Accession # P01034 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Cystatin C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatins A, B, D, E/M, S, SA, SN, rhFetuins A, B, or rhHPRG (Histidine-Proline-Rich Glycoprotein) is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title