missing translation for 'onlineSavingsMsg'
Learn More

Human Cytokeratin 17 Antibody, R&D Systems™

Mouse Monoclonal Antibody

247.00 € - 536.00 €

Specifications

Antigen Cytokeratin 17
Clone 1080235
Dilution Western Blot 2 μg/mL, Simple Western 100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
Applications Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30245517
View Documents
R&D Systems
MAB11571-SP
25 μg
247.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30245516
View Documents
R&D Systems
MAB11571-100
100 μg
536.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for Immunocytochemistry,Western Blot,Simple Western,Immunohistochemistry
Specifications

Specifications

Cytokeratin 17
Western Blot 2 μg/mL, Simple Western 100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin 17, type I, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1
Cytokeratin 17 containing synthetic peptide, Accession # Q04695
Primary
Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
1080235
Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry
Unconjugated
Mouse
Human
Q04695
3872
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.