missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytokeratin 19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB35063-SP
This item is not returnable.
View return policy
Description
Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Specifications
| Cytokeratin 19 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, EndoC, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, Krt19, MGC15366 | |
| E. coli-derived human Cytokeratin 19, Gln311-Lys370, Accession # P08727 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 1042608 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
| P08727 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3880 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction