missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dectin-1/CLEC7A PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
278.00 € - 492.00 €
Specifications
| Antigen | Dectin-1/CLEC7A |
|---|---|
| Clone | 259931 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765357
![]() |
R&D Systems
FAB1859P-025 |
25 Tests |
278.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18798206
![]() |
R&D Systems
FAB1859P-100 |
100 Tests |
492.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Human samples. It is validated for Flow Cytometry.Specifications
| Dectin-1/CLEC7A | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| CLEC7A | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Dectin-1/CLEC7A in direct ELISAs. In directs ELISAs, less than 10% cross-reactivity with recombinant human (rh) DLEC and no cross-reactivity with recombinant mouse Dectin-1 or rhDectin-2 is observed. |
| 259931 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| NP_072092 | |
| 64581 | |
| Mouse myeloma cell line NS0-derived recombinant human Dectin-1/CLEC7A Thr66-Met201 Accession # NP_072092 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title