Learn More
Human Desmoplakin Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB9080
 View more versions of this product
Product Code. 18710588
  • 429.00 € / 100µg
Description
Desmoplakin Monoclonal specifically detects Desmoplakin in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Desmoplakin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
250/210 kDa paraneoplastic pemphigus antigen, DCWHKTA, desmoplakin, desmoplakin (DPI, DPII), desmoplakin I, desmoplakin II, DP, DPI, DPII, DSP, KPPS2, PPKS2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
1832
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot
824038
Western Blot 0.5 ug/mL
P15924
DSP
E. coli-derived recombinant human Desmoplakin Asn1796-Val2046 Accession # P15924
100 μg
Primary
Detects human Desmoplakin in direct ELISAs and Western blots.
Human
Purified